LIFH324A10CD Life Chemicals Inc.

1-(2-methyl-1-benzothiophen-3-yl)methanamine hydrochloride

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart