LIFH324A10EE Life Chemicals Inc.

2-chloro-5H,6H,7H-pyrrolo[1,2-a]imidazole

  • MDL number MFCD01563751
  • Cas Number : 91417-91-3
  • Molecular Weight : 142.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart