LIFH4466608B Life Chemicals Inc.

3-azabicyclo[3.2.1]octane hydrochloride

  • MDL number MFCD22196503
  • Cas Number : 20969-02-2
  • Molecular Weight : 147.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart