LIFH44666098 Life Chemicals Inc.

2,2-dimethylhex-5-en-1-ol

  • MDL number MFCD16038666
  • Cas Number : 56068-50-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart