LIFH4466609C Life Chemicals Inc.

6-phenylpyrimidine-4-carbonitrile

  • MDL number MFCD00234690
  • Cas Number : 83858-06-4
  • Molecular Weight : 181.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart