LIFH446660B7 Life Chemicals Inc.

4,7-dimethoxy-3-methyl-2,3-dihydro-1,3-benzothiazol-2-imine; sulfuric acid

  • MDL number MFCD32174368
  • Cas Number : 2379946-27-5
  • Molecular Weight : 322.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart