LIFH446660D5 Life Chemicals Inc.

3-fluoro-4-methoxy-N-methylbenzamide

  • MDL number MFCD11743296
  • Cas Number : 633317-76-7
  • Molecular Weight : 183.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart