LIFH446660FC Life Chemicals Inc.

tert-butyl N-[4-(2-hydroxyethyl)phenyl]carbamate

  • MDL number MFCD04974330
  • Cas Number : 104060-23-3
  • Molecular Weight : 237.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart