LIFH44666103 Life Chemicals Inc.

4-(3-{[(tert-butoxy)carbonyl]amino}propoxy)benzoic acid

  • MDL number MFCD20485942
  • Cas Number : 174664-94-9
  • Molecular Weight : 295.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-{3-[(tert-butoxycarbonyl)amino]propoxy}benzoic acid
Inchi Code 1S/C15H21NO5/c1-15(2,3)21-14(19)16-9-4-10-20-12-7-5-11(6-8-12)13(17)18/h5-8H,4,9-10H2,1-3H3,(H,16,19)(H,17,18)
Inchi Key WBPCCYDKFTVWOK-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart