LIFH55F4F2AE Life Chemicals Inc.

[5-(aminomethyl)thiophen-2-yl](phenyl)methanol

  • MDL number MFCD24387835
  • Cas Number : 1447963-63-4
  • Molecular Weight : 219.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart