LIFH55F4F2BF Life Chemicals Inc.

3-methyl-1-(propan-2-yl)-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD08700655
  • Cas Number : 50920-68-8
  • Molecular Weight : 168.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-isopropyl-3-methyl-1H-pyrazole-5-carboxylic acid
Inchi Code 1S/C8H12N2O2/c1-5(2)10-7(8(11)12)4-6(3)9-10/h4-5H,1-3H3,(H,11,12)
Inchi Key AUZHMOQAPWTGTB-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart