LIFH55F4F2D2 Life Chemicals Inc.

3-[(piperidin-4-yl)methyl]-3,4-dihydroquinazolin-4-one dihydrochloride

  • Cas Number : 2379946-35-5
  • Molecular Weight : 316.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart