LIFH55F4F31A Life Chemicals Inc.

1-[(azetidin-3-yl)methyl]-4-chloro-1H-pyrazole hydrochloride

  • Cas Number : 2379945-52-3
  • Molecular Weight : 208.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart