LIFH55F4F32E Life Chemicals Inc.

1-(4-tert-butylphenyl)-6,6-dimethyl-4,5,6,7-tetrahydro-1H-indazol-4-one

  • Cas Number : 2379946-55-9
  • Molecular Weight : 296.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart