LIFH55F4F332 Life Chemicals Inc.

tert-butyl N-{3-[(4-formylnaphthalen-1-yl)oxy]propyl}carbamate

  • Cas Number : 1184952-24-6
  • Molecular Weight : 329.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart