LIFH580D634C Life Chemicals Inc.

5-(3,3-difluorocyclobutyl)imidazolidine-2,4-dione

  • Cas Number : 2379945-26-1
  • Molecular Weight : 190.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(3,3-difluorocyclobutyl)-1H-imidazole-2,4-diol

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart