LIFH580D634D Life Chemicals Inc.

ethyl 2-fluoropent-4-ynoate

  • Cas Number : 2378806-54-1
  • Molecular Weight : 144.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart