LIFH580D6350 Life Chemicals Inc.

2-iodo-4,6-dimethylpyridin-3-amine

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart