LIFH580D6351 Life Chemicals Inc.

3,5-dibromo-4-iodopyridin-2-ol

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart