LIFH580D6352 Life Chemicals Inc.

2-iodo-4,6-dimethylpyridine-3-carboxylic acid

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart