LIFH580D6357 Life Chemicals Inc.

methyl 4-bromo-3-(trifluoromethyl)benzoate

  • MDL number MFCD10566409
  • Cas Number : 107317-58-8
  • Molecular Weight : 283.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart