LIFH580D6358 Life Chemicals Inc.

tert-butyl 3-bromo-5-(trifluoromethyl)benzoate

  • MDL number MFCD16037034
  • Cas Number : 1237535-81-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart