LIFH580D635A Life Chemicals Inc.

1-(6-chloropyridin-3-yl)-2-methylpropan-1-one

  • MDL number MFCD08437179
  • Cas Number : 244263-45-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart