LIFH580D635B Life Chemicals Inc.

tert-butyl 4-(2,2,2-trifluoro-1-hydroxyethyl)piperidine-1-carboxylate

  • MDL number MFCD11501657
  • Cas Number : 184042-83-9
  • Molecular Weight : 283.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart