LIFH580D635C Life Chemicals Inc.

1-benzyl 5-tert-butyl (5S)-2-oxoimidazolidine-1,5-dicarboxylate

  • MDL number MFCD11046510
  • Cas Number : 77999-24-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart