LIFH580D635D Life Chemicals Inc.

1-(4-{2-azabicyclo[2.2.1]heptan-2-yl}phenyl)methanamine

  • MDL number MFCD14696267

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart