LIFH580D635F Life Chemicals Inc.

3-bromo-5-formylbenzonitrile

  • MDL number MFCD09261040
  • Cas Number : 644982-55-8
  • Molecular Weight : 210.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-5-formylbenzonitrile
Inchi Code 1S/C8H4BrNO/c9-8-2-6(4-10)1-7(3-8)5-11/h1-3,5H
Inchi Key FKGQJUAOTNPVFD-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart