LIFH580D6360 Life Chemicals Inc.

6-(2,2,2-trifluoroethoxy)pyrimidine-4-carboxylic acid

  • MDL number MFCD26096822
  • Cas Number : 1439896-74-8
  • Molecular Weight : 222.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart