LIFH580D6361 Life Chemicals Inc.

2-[1-(2-methoxyethyl)-1H-pyrazol-4-yl]ethan-1-amine

  • MDL number MFCD16862518
  • Cas Number : 1343369-80-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart