LIFH580D6364 Life Chemicals Inc.

2,2-bis(2-bromoethyl)-1,3-dioxolane

  • MDL number MFCD09833653
  • Cas Number : 164987-79-5
  • Molecular Weight : 287.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,2-bis(2-bromoethyl)-1,3-dioxolane
Inchi Code 1S/C7H12Br2O2/c8-3-1-7(2-4-9)10-5-6-11-7/h1-6H2
Inchi Key NXMURIASTYIOQB-UHFFFAOYSA-N

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart