LIFH580D6366 Life Chemicals Inc.

bis(4-hydroxy-5-methoxypyridine-2-carboxylic acid) hydrate

  • Cas Number : 2378806-56-3
  • Molecular Weight : 356.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-hydroxy-5-methoxypicolinic acid hemihydrate

Description

Purity 95%
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart