LIFH580D6368 Life Chemicals Inc.

3-(4-aminophenyl)propanenitrile

  • MDL number MFCD09836148
  • Cas Number : 10139-91-0
  • Molecular Weight : 146.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart