LIFH580D6374 Life Chemicals Inc.

methyl 3-benzyl-3-azabicyclo[3.2.0]heptane-1-carboxylate

  • MDL number MFCD29999890
  • Cas Number : 2098137-34-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart