LIFH580D6378 Life Chemicals Inc.

5-(6-aminopyridin-3-yl)thiophene-2-carboxylic acid hydrate

  • Cas Number : 2378806-60-9
  • Molecular Weight : 238.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart