LIFH580E1937 Life Chemicals Inc.

5-iodo-6-methyl-1,7-naphthyridine

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart