LIFH580E1938 Life Chemicals Inc.

bis(N-(pyridin-2-yl)pyrrolidine-3-carboxamide) trihydrochloride

  • Cas Number : 2379945-77-2
  • Molecular Weight : 491.85

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart