LIFH580E194B Life Chemicals Inc.

4,8-dimethylnon-7-enenitrile

  • Cas Number : 52671-32-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart