LIFH580E194C Life Chemicals Inc.

5,11-dihydroxy-1,7-diazatricyclo[7.3.0.0^{3,7}]dodecane-2,8-dione

  • Cas Number : 94292-34-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart