LIFH580E194E Life Chemicals Inc.

tert-butyl 3-(carbamoylamino)piperidine-1-carboxylate

  • MDL number MFCD24394168

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart