LIFH580E195B Life Chemicals Inc.

6,6-difluorobicyclo[3.1.0]hexan-2-one

  • MDL number MFCD22548269
  • Cas Number : 1393576-51-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart