LIFH580E195C Life Chemicals Inc.

6-phenylspiro[3.3]heptane-2-carboxylic acid

  • MDL number MFCD29046423

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart