LIFH580E195E Life Chemicals Inc.

1-bromo-3,3-dimethylbutan-2-ol

  • MDL number MFCD03844392
  • Cas Number : 1438-13-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart