LIFH580E1971 Life Chemicals Inc.

(2,2-dimethylpropyl)[(pyrazin-2-yl)methyl]amine

  • MDL number MFCD16864734

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart