LIFH580E197B Life Chemicals Inc.

1-[1-(pyridin-3-yl)-1H-1,2,3-triazol-4-yl]methanamine

  • MDL number MFCD26620871

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart