LIFH5F6BEB23 Life Chemicals Inc.

5-bromo-4-methyl-2-(propan-2-yl)-1,3-thiazole

  • MDL number MFCD22031307
  • Cas Number : 910552-70-4
  • Molecular Weight : 220.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart