LIFH5F6BEB52 Life Chemicals Inc.

1-(5-methyl-1,3,4-thiadiazol-2-yl)pyrrolidin-3-ol

  • MDL number MFCD21182401
  • Cas Number : 1487131-19-0
  • Molecular Weight : 185.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart