LIFH89538898 Life Chemicals Inc.

1-(3,4-dihydro-1H-2-benzopyran-3-yl)methanamine hydrochloride

  • Cas Number : 2413376-14-2
  • Molecular Weight : 199.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 91
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart