LIFH895388A1 Life Chemicals Inc.

2-(trifluoromethyl)-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD16616651
  • Cas Number : 147642-16-8
  • Molecular Weight : 179.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart