LIFH895388AD Life Chemicals Inc.

3-bromo-5-(2,2-difluorocyclopropyl)benzonitrile

  • Cas Number : 2413374-02-2
  • Molecular Weight : 258.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart