LIFH93CB08D2 Life Chemicals Inc.

N-cyclopropyl-2,6-dimethylpyrimidin-4-amine

  • MDL number MFCD12798488
  • Cas Number : 1178628-52-8
  • Molecular Weight : 163.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-cyclopropyl-2,6-dimethyl-4-pyrimidinamine
Inchi Code 1S/C9H13N3/c1-6-5-9(11-7(2)10-6)12-8-3-4-8/h5,8H,3-4H2,1-2H3,(H,10,11,12)
Inchi Key LCKJMDYLGJSBBC-UHFFFAOYSA-N

Description

Purity 95%
Physical Form oil
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart